Anti-FGF18 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX128496
10166-796EA 577.14 USD
10166-796
Anti-FGF18 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to FGF18 (fibroblast growth factor 18)

Type: Primary
Antigen: FGF18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR