Anti-IgNar Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX128444
10793-272EA 983.64 USD
10793-272
Anti-IgNar Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Horn shark IgNar

Type: Primary
Antigen: IgNar
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Shrk
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR