Anti-prox1a Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX128354
10167-186EA 618.01 USD
10167-186
Anti-prox1a Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Prox1 (prospero-related homeobox gene 1a)

Type: Primary
Antigen: prox1a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR