Anti-myl7 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX128346
10167-184EA 588.27 USD
10167-184
Anti-myl7 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Myl7 (myosin, light polypeptide 7, regulatory)

Type: Primary
Antigen: myl7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebra Fish, Medaka
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR