Anti-PCV Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX128121
89424-576EA 582.76 USD
89424-576
Anti-PCV Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Porcine circovirus type 2 Capsid Protein

Type: Primary
Antigen: PCV2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Porcine circovirus type 2
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR