Anti-GJA5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX128055
10166-760EA 577.14 USD
10166-760
Anti-GJA5 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Connexin 40 (gap junction protein, alpha 5, 40kDa)

Type: Primary
Antigen: GJA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR