Anti-IKZF4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX128043
10166-758EA 557.62 USD
10166-758
Anti-IKZF4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to IKZF4 (IKAROS family zinc finger 4 (Eos))

Type: Primary
Antigen: IKZF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR