Anti-PREX1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX128035
89424-532EA 582.76 USD
89424-532
Anti-PREX1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PREX1 (phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 1)

Type: Primary
Antigen: PREX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR