Anti-CEP152 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX128027
89424-524EA 582.76 USD
89424-524
Anti-CEP152 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CEP152 (centrosomal protein 152kDa)

Type: Primary
Antigen: CEP152
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR