Anti-C. koseri Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12796
10165-680EA 445.47 USD
10165-680
Anti-C. koseri Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to Citrobacter koseri

Type: Primary
Antigen: C. koseri
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR