Anti-Gap43 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX127937
89424-480EA 617.09 USD
89424-480
Anti-Gap43 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to GAP43 (growth associated protein 43)

Type: Primary
Antigen: Gap43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR