Anti-SETD2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX127906
89424-462EA 582.76 USD
89424-462
Anti-SETD2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to HYPB/SETD2 (SET domain containing 2)

Type: Primary
Antigen: SETD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR