Anti-h2afx Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX127342
89424-450EA 638.39 USD
89424-450
Anti-h2afx Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to zebrafish histone H2A.X (phospho Ser139) (H2A histone family, member X)

Type: Primary
Antigen: h2afx
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR