Anti-H9N2 HA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX127307
89424-416EA 582.76 USD
89424-416
Anti-H9N2 HA Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Influenza A Virus H9N2 Hemagglutinin (HA)

Type: Primary
Antigen: H9N2 HA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Influenza A virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR