Anti-H1N1 HA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX127294
89424-402EA 488.2 USD
89424-402
Anti-H1N1 HA Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Influenza A Virus H1N1 Hemagglutinin (HA)

Type: Primary
Antigen: H1N1 HA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Influenza A virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR