Anti-cdh2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX125962
10167-170EA 617.99 USD
10167-170
Anti-cdh2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to N-cadherin (cadherin 2, neuronal)

Type: Primary
Antigen: cdh2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR