Anti-SLU7 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX125955
89424-384EA 582.76 USD
89424-384
Anti-SLU7 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Slu7 (SLU7 splicing factor homolog (S. cerevisiae))

Type: Primary
Antigen: SLU7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR