Anti-USP7 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX125894
10166-740EA 577.14 USD
10166-740
Anti-USP7 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to USP7 (ubiquitin specific peptidase 7 (herpes virus-associated))

Type: Primary
Antigen: USP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR