Anti-Tyramine Rat Polyclonal Antibody

GTX12578
10165-660EA 404.65 USD
10165-660
Anti-Tyramine Rat Polyclonal Antibody
Rat Polyclonal antibody to Tyramine

Type: Primary
Antigen: Tyramine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Other
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR