Anti-DAG1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX124225
89424-366EA 615.35 USD
89424-366
Anti-DAG1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to beta dystroglycan (dystroglycan 1 (dystrophin-associated glycoprotein 1))

Type: Primary
Antigen: DAG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR