Anti-LAMA2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12391
10162-510EA 628.2 USD
10162-510
Anti-LAMA2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to LAMA2

Type: Primary
Antigen: LAMA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR