Anti-ARID1A Rabbit Polyclonal Antibody

GTX12388
10162-506EA 442.38 USD
10162-506
Anti-ARID1A Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to ARID1A

Type: Primary
Antigen: ARID1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR