Anti-SPINK1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX123685
10166-380EA 573.94 USD
10166-380
Anti-SPINK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SPINK1 (serine peptidase inhibitor, Kazal type 1)

Type: Primary
Antigen: SPINK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR