Anti-SPATA4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX123559
89424-340EA 582.76 USD
89424-340
Anti-SPATA4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TSARG2 (spermatogenesis associated 4)

Type: Primary
Antigen: SPATA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR