Anti-HNRNPDL Rabbit Polyclonal Antibody

GTX123497
10166-730EA 400.11 USD
10166-730
Anti-HNRNPDL Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to HNRPDL (heterogeneous nuclear ribonucleoprotein D-like)

Type: Primary
Antigen: HNRNPDL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR