Anti-Prlr Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12307
10165-446EA 643.6 USD
10165-446
Anti-Prlr Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PRLR

Type: Primary
Antigen: Prlr
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR