Anti-MADCAM1 Rabbit Polyclonal Antibody

GTX12305
10165-442EA 442.38 USD
10165-442
Anti-MADCAM1 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to MADCAM1

Type: Primary
Antigen: MADCAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR