Anti-SCLY Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12283
10165-416EA 628.2 USD
10165-416
Anti-SCLY Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SCLY

Type: Primary
Antigen: SCLY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR