Anti-TRPV6 Rabbit Polyclonal Antibody

GTX12280
10165-410EA 442 USD
10165-410
Anti-TRPV6 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to TRPV6

Type: Primary
Antigen: TRPV6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR