Anti-TJP3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12279
10165-408EA 643.93 USD
10165-408
Anti-TJP3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TJP3

Type: Primary
Antigen: TJP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR