Anti-FSTL3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12198
10165-380EA 628.2 USD
10165-380
Anti-FSTL3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to FSTL3

Type: Primary
Antigen: FSTL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR