Anti-Cyp2e1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12194
10165-376EA 643.6 USD
10165-376
Anti-Cyp2e1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CYP2E1

Type: Primary
Antigen: Cyp2e1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR