Anti-Tfr2 Rabbit Polyclonal Antibody

GTX12158
10165-372EA 442.38 USD
10165-372
Anti-Tfr2 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to TFR2

Type: Primary
Antigen: Tfr2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR