    Sort by:
  • Search Within Results
gensolve-310
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (89400-778)
Supplier:  KMG
Description:   The Gensolve product line are organic stripping compositions that remove hydrophillic polymeric organic coating, such as photoresists and their residu...