    Sort by:
  • Search Within Results
pad-etch-4
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (89400-808)
Supplier:  KMG
Description:   Pad etch is a gentle silicon dioxide etchant that minimizes corrosion on underlying aluminum.