    Sort by:
You Searched For:
CR-16 Chrome etch
1  results were found
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (10803-036)
Supplier:  KMG
Description:   CR-16, is a nitric acid based chrome etchant used for photolithography masks, semiconductor and MEMS processes. It is designed to provide minimum unde...