    Sort by:
  • Search Within Results
Buffered oxide etch 50:1
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (89400-756)
Supplier:  KMG
Description:   Buffered oxide etchants are used to etch thin films of silicon dioxide and shape contact and via openings.