tert-Butyldimethyl(tributylstannylmethoxy)silane 90-95%

Supplier: Matrix Scientific

115969-5G
102900-050EA 884.62 USD
102900-050
tert-Butyldimethyl(tributylstannylmethoxy)silane 90-95%
tert-Butyldimethyl(tributylstannylmethoxy)silane

Hazard: Irritant/Refrigerate

Formula: C₁₉H₄₄OSiSn
MW: 435.34 g/mol
MDL Number: MFCD23152698
CAS Number: 123061-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR