2-Benzyl-2,7-diaza-spiro[4.4]nonane

Supplier: Matrix Scientific

Synonyms: 2-Benzyl-2,7-diazaspiro[4.4]nonane

113105-1G
102891-494EA 1018.46 USD
102891-494
2-Benzyl-2,7-diaza-spiro[4.4]nonane
2-Benzyl-2,7-diaza-spiro[4.4]nonane

Hazard: Irritant

Formula: C₁₄H₂₀N₂
MW: 216.33 g/mol
MDL Number: MFCD04115133
CAS Number: 885275-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR