Cyclohexylacetyl chloride ≥97%

Supplier: Matrix Scientific

114594-1G
102894-360EA 183.08 USD
102894-360
Cyclohexylacetyl chloride ≥97%
Cyclohexylacetyl chloride

Hazard: Corrosive

Formula: C₈H₁₃ClO
MDL Number: MFCD03424702
CAS Number: 23860-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR