10,11-Dihydro-5H-dibenzo[a,d]cycloheptene-5-methanol ≥95%

Supplier: Matrix Scientific

115772-500MG 115772-1G
102900-260EA 1209.23 USD
102900-260 102899-656
10,11-Dihydro-5H-dibenzo[a,d]cycloheptene-5-methanol ≥95%
10,11-Dihydro-5H-dibenzo[a,d]cycloheptene-5-methanol

Hazard: Irritant

Formula: C₁₆H₁₆O
MDL Number: MFCD22573382
CAS Number: 2975-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR