Methyl-6-bromo-2-oxo-1,2,3,4-tetrahydro-1,8-naphthyridine-3-carboxylate ≥95%
Supplier: Matrix Scientific
115722-10G
115722-5G
115722-1G
102899-482EA
743.08
USD
102899-482
102899-486
102899-484
Methyl-6-bromo-2-oxo-1,2,3,4-tetrahydro-1,8-naphthyridine-3-carboxylate ≥95%
Methyl-6-bromo-2-oxo-1,2,3,4-tetrahydro-1,8-naphthyridine-3-carboxylate
Hazard: Irritant
Formula:
C₁₀H₉BrN₂O₃ |
MDL Number:
MFCD08275095 CAS Number: 335031-10-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...