(S)-1-Methyl-3-aminomethylpiperidine ≥95%

Supplier: Matrix Scientific

115642-1G
102897-042EA 1373.85 USD
102897-042
(S)-1-Methyl-3-aminomethylpiperidine ≥95%
(S)-1-Methyl-3-aminomethylpiperidine

Hazard: Irritant

Formula: C₇H₁₆N₂
MDL Number: MFCD11226132
CAS Number: 1217604-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR