2-Bromo-(5-(1-methyl-4-piperidinyl)methoxy)-2-pyrimidine ≥95%

Supplier: Matrix Scientific

115623-1G
102897-088EA 2721.54 USD
102897-088
2-Bromo-(5-(1-methyl-4-piperidinyl)methoxy)-2-pyrimidine ≥95%
2-Bromo-(5-(1-methyl-4-piperidinyl)methoxy)-2-pyrimidine

Hazard: Irritant

Formula: C₁₁H₁₆BrN₃O
MDL Number: MFCD23105889
CAS Number: 1400764-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR