9-Cbz-2-oxo-3-oxa-1,9-diazaspiro[5.5]undecane ≥95%
Supplier: Matrix Scientific
115601-500MG
115601-250MG
102896-986EA
1964.62
USD
102896-986
102896-988
9-Cbz-2-oxo-3-oxa-1,9-diazaspiro[5.5]undecane ≥95%
9-Cbz-2-oxo-3-oxa-1,9-diazaspiro[5.5]undecane
Hazard: Irritant
Formula:
C₁₆H₂₀N₂O₄ MW: 304.35 g/mol |
MDL Number:
MFCD22415196 CAS Number: 1363382-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...