8-Boc-2-oxo-3-oxa-1,8-diazaspiro[5.5]undecane ≥95%

Supplier: Matrix Scientific

115600-500MG
102896-696EA 3295.38 USD
102896-696
8-Boc-2-oxo-3-oxa-1,8-diazaspiro[5.5]undecane ≥95%
8-Boc-2-oxo-3-oxa-1,8-diazaspiro[5.5]undecane

Hazard: Irritant

Formula: C₁₃H₂₂N₂O₄
MW: 270.33 g/mol
MDL Number: MFCD22415208
CAS Number: 1363381-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR