1-[(1-Chlorocyclobutyl)sulfinyl]-4-methylbenzene ≥95%

Supplier: Matrix Scientific

115537-100MG 115537-250MG
102896-744EA 1318.46 USD
102896-744 102896-746
1-[(1-Chlorocyclobutyl)sulfinyl]-4-methylbenzene ≥95%
1-[(1-Chlorocyclobutyl)sulfinyl]-4-methylbenzene

Hazard: Irritant

Formula: C₁₁H₁₃ClOS
MDL Number: MFCD22370066
CAS Number: 1221171-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR