4,5-Dihydro-2H-benzo[g]indazole-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

115093-1G 115093-500MG
102891-588EA 1040 USD
102891-588 102891-586
4,5-Dihydro-2H-benzo[g]indazole-3-carboxylic acid ≥97%
4,5-Dihydro-2H-benzo[g]indazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₂
MDL Number: MFCD08733223
CAS Number: 898796-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR