2-(4-Isopropylphenyl)pyrazolo[1,5-a]pyrazin-4(5H)-one ≥95%
Supplier: Matrix Scientific
115068-500MG
115068-1G
102895-344EA
1040
USD
102895-344
102895-342
2-(4-Isopropylphenyl)pyrazolo[1,5-a]pyrazin-4(5H)-one ≥95%
2-(4-Isopropylphenyl)pyrazolo[1,5-a]pyrazin-4(5H)-one
Hazard: Irritant
Formula:
C₁₅H₁₅N₃O MW: 253.31 g/mol |
MDL Number:
MFCD25951875 CAS Number: 1338666-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...