4-(3-tert-Butyl-1,2,4-oxadiazol-5-yl)-2-chloropyridine ≥97%
Supplier: Matrix Scientific
115048-500MG
115048-1G
102895-288EA
1040
USD
102895-288
102895-286
4-(3-tert-Butyl-1,2,4-oxadiazol-5-yl)-2-chloropyridine ≥97%
4-(3-tert-Butyl-1,2,4-oxadiazol-5-yl)-2-chloropyridine
Hazard: Irritant
Formula:
C₁₁H₁₂ClN₃O |
MDL Number:
MFCD20528957 CAS Number: 1338675-20-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...